- Description:Benchtop Water Quality Meter (pH, Conductivity, Salinity, TDS, Temperature)
- Dimensions:217 × 168 × 58 mm (8.5 × 6.5 × 2.25")
- Weight:533 g (18 oz.)
- Cat. no.:470204-194
- Supplier No.:860033
Reads pH, ORP (oxygen reduction potential), mV, conductivity, TDS (total dissolved solids), salinity and temperature.
- Durable Housing
- Easy to Read
- Automatic or manual temperature compensation
- Automatic or manual ranging, automatic buffer recognition
- Digital and analog outputs
- Min/max and 99 data points, data hold
- 5 calibration points
- Electrode status indication
Operates on AC voltage (no batteries). Also accepts any pH or ORP probe with a standard BNC connector.
Rugged housing with large LCD displaying the parameter being measured together with time, date and temperature (in °C or °F). Measures: ph, ORP (oxygen reduction potential - SOLD SEPARATELY), mV, Conductivity, TDS (total dissolved solids), Salinity and Temperature.
pH Range: 0 - 14. Resolution: 0.01pH. Accuracy: ±0.02pH.
Conductivity Range: 0 - 19.9, 199.9, 1999 μS/cm; 0 - 19.9, 199.9 mS/cm. Resolution: 0.01, 0.1, 1μS/cm; 0.01, 0.1mS/cm. Accuracy: ±1% fs + 1d
TDS Range: 0 - 19.9, 199.9, 1999 ppm; 0 - 19.9, 199. 9 ppt. Resolution: 0.01, 0.1, 1ppm; 0.01, 0.1ppt. Accuracy: ±1% fs + 1d
Salinity Range: 0 - 11.38, 80 ppt. Resolution: 0.01, 0.1ppt. Accuracy: ±1% fs + 1d.
Temperature Range: -5°C - 80C; 23 - 176°F. Resolution: 0.1°F/°C. Accuracy: ±0.9°F/0.5°C