Order Entry
Canada
ContactUsLinkComponent
Beaker Tongs
Beaker Tongs
Catalog # 470019-542
undefined
Beaker Tongs
Catalog # 470019-542
Supplier Number:  BTSP09

Specifications

  • Description:
    Beaker Tongs
  • Cat. no.:
    470019-542
  • Supplier No.:
    H-23422

Specifications

About this item

Equipped with convenient plastic coated jaws.

  • Zinc-plated steel
  • Opens to 7"
  • Overall length is 9.75"

These beaker tongs are constructed with flat riveted joints and convenient vinyl coated sleeves. Tongs can handle beakers ranging from 50 to 1500 mL capacity.