Basic Protist Survey Slide Set
Basic Protist Survey Slide Set, 6, Protists
Catalog # 470183-454
Supplier: Avantor
Jump to:
Product Details & Documents
Representatives from four Phyla
- Identifying features clearly distinguishable
- Rigorous quality control standards
All slides are prepared as whole mounts.
Ordering information: Set includes Chlorophyta: Spirogyra, vegetative; Chrysophyta (Stramenopila): Diatoms, mixed; Ciliata (Alveolata): Paramecium; Euglenozoa: Euglena; Rhodophyta: Bactrachospermum; and Sarcodina (Rhizopod): Amoeba.
Specifications
- Type:Protists
- Description:Basic Protist Survey Slide Set
- No. of slides:6