BACTIDENT™ Rapid Tests: Indole and Oxidase Reagents
Supplier: MilliporeSigma
Certificates
About this item
Indole Reagent
Indole Reagent for the Indole reaction of Enterobacteria. 30 ml bottle.
Oxidase Test Strips
Test Strips to detect production of cytochrome oxidase in microorganisms. Package of 50 strips.
Reagents ship at ambient temperature. Store at +2°C to +8°C.
Frequently Bought Together