You searched for: Chemical Fume Hoods
Designed to guarantee laboratory conditions and researchers remain safe at all times, the chemical fume hoods protect against flammable or toxic chemicals. These protection devices have clear sashes that slide horizontally or vertically into place and support high working visibility. Constructed for maximum chemical resistance, the leak-proof enclosures will not corrode even with heavy routine use and corrosive spills. The chemical fume hoods circulate and filter air to eliminate possible vapors produced.
VWR® REDISHIP General Purpose Fume Hoods with Cupsink, Kewaunee Scientific
Supplier: VWR International
General purpose fume hoods that meet the typical needs of a research environment, while maintaining a level of flexibility. Ready for quick delivery through the REDISHIP program.
Expand 3 Items
REDISHIP Supreme Air® Fittings for KMER-Lined Bench and Walk-In Hoods, Kewaunee®
Supplier: VWR International
These remote control fittings consist of a brass angle valve with replaceable seat, a nylon color-coded panel flange with angle serrated hose connector (except VFT-0864-3WV, which features a gooseneck), a remote control rod assembly with a plastic color-coded four-arm handle, black plastic angle flange, and a tank nipple, locknut, and washer
Expand 7 Items
REDISHIP Protector® Premier® Laboratory Hoods and REDISHIP SpillStopper™ Work Surfaces, Labconco®
Supplier: Labconco
At 100 fpm and sas fully open, uses 730 CFM. Powder-coated steel exterior. Molded one piece fiberglass liner. By-pass block. 37.5inch sightline. Sash stop at 18inch. L and R GFCI. EA.
Expand 4 Items
Protector® Fume Hood Airflow Monitors, Service Fixture Kits, and Duplex Receptacle Kits, Labconco
Supplier: Labconco
There are many accessories available for customizing Protector® Laboratory Hoods including Service Fixture Kits, Duplex Receptacle Kits and Guardian™ Airflow Monitors.
Expand 20 Items
REDISHIP Protector® XStream® Laboratory Hoods Customisable with REDISHIP SpillStopper™ Work Surfaces, Labconco®
Supplier: Labconco
At 60 fpm and sash fully open, uses only 425 CFM. Powder-coated steel exterior. Composite panel liner. 37.5inch high unobstructed sightline. L and R GFCI. . EA.
Expand 4 Items
Guardian™ Airflow Monitor Kits, Labconco®
Supplier: Labconco
Guardian™ airflow monitors continuously check the face velocity of laboratory hoods and enclosures and alert the operator to airflows that drop outside preset levels with audible and visual alarms. The audible alarm can be muted at the touch of a switch.
Expand 10 Items
Adjustable Height Base Stands, Labconco®
Supplier: Labconco
For use with Basic™ 47, Basic 70, Fiberglass 30, Protector® XStream®, Protector Premier™, Protector ClassMate®, Protector PVC Perchloric Acid, Protector Stainless Steel Perchloric Acid, Logic+, Logic, Cell Logic, PuriCare Procedure Stations and Protector Radioisotope laboratory hoods
Expand 15 Items
SP Bel-Art Fabricated Fume Hood, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
The translucent polypropylene plastic hood is non-conductive, chemical-resistant and corrosion-proof.
Expand 1 Items
SP Bel-Art Large Molded Fume Hood with Baffle, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
This fume hood includes a flat internal baffle across the rear of the shell that slopes forward, leaving space beneath for air flow to sweep the deck, and space above in order to efficiently remove airborne vapors.
Expand 1 Items
SP Bel-Art Clear View Fume Hoods, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Safely vent noxious or potentially hazardous fumes and dust using Scienceware® Clear View Fume Hoods.
Expand 2 Items
SP Bel-Art Large Molded Fume Hood, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
This fume hood features a one-piece shell molded of polyethylene plastic with a clear acrylic sash that slides on inside tracks.
Expand 1 Items
SP Bel-Art Tapered Rear Exhaust Fume Hoods, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
These corrosion-proof polypropylene plastic exhaust hoods are designed to rest on the rear edge of tanks and draw a stream of air across the top of the tank away from the operator.
Expand 1 Items
Fiberglass 30 Laboratory Hoods, Labconco®
Supplier: Labconco
For use with a remote blower. Includes pre-wired incandescent light and blower switches. Vertical-rising, 0.5cm thick sash. 115V, 60Hz. Exterior dimensions: 76.2Wx76.5Lx121.9Hcm. Maximum static pressure loss: 0.12" H2O at 100fpm. Shipping weight: 43.6kg. UL listed.
Expand 6 Items
Distillation Grid Kits for Protector®, Premier™ and XStream® Fume Hoods, Labconco®
Supplier: Labconco
These kits let the user mount glassware and other small apparatus to the rear of the hood’s interior, above the work surface to maximize usable working area.
Expand 4 Items
Protector® Premier® I-S™ Laboratory Fume Hoods, Labconco Corporation
Supplier: Labconco
High performance fume hoods with integrated sash sensors for automatic multi-speed blower control.
Expand 8 Items
REDISHIP Air Alert 300 Face Velocity Airflow Alarm, Kewaunee®
Supplier: Kewaunee
Air Alert 300 Face Velocity Airflow Alarm consists of a thermistor sensor mounted through the end wall of the hood, and a control monitor that gives both a visual and an audible alarm.
Expand 1 Items
Protector® XStream® I-S™ Laboratory Fume Hoods, Labconco
Supplier: Labconco
High performance fume hoods with integrated sash sensors for automatic multi-speed blower control.
Expand 8 Items
Solid Epoxy Flat Work Surfaces, Labconco®
Supplier: Labconco
Combine with a base stand or cabinet to support Fiberglass 30 and Basic™ Labconco® laboratory hoods.
Expand 6 Items
Protector® XStream® Laboratory Hoods, Labconco®
Supplier: Labconco
The newly redesigned, patented Protector® XStream® Laboratory Hood was engineered to be the best containing fume hood possible
Expand 16 Items
Venturi V05-General Purpose Bench Fume Hood with Vertical Rising Sash, Kemglass Liner, Kewaunee Scientific
Supplier: Kewaunee
The Supreme Air Venturi provides the safest user-friendly fume hood manufactured today. The Supreme Air Venturi utilizes proven containment enhancing techniques coupled with innovative laboratory convenience features, including: adjustable LED lights, electromechanical sash stops, ultra-smooth sash system, and push button sash management.
Expand 1 Items
Protector® Radioisotope Laboratory Hoods, Labconco®
Supplier: Labconco
Designed to give maximum protection for radiochemical applications.
Expand 40 Items
Electrical Receptacle Kits for Basic™ Laboratory Hoods, Labconco®
Supplier: Labconco
Kits include electrical receptacle, junction box, receptacle cover plate, and installation instructions
Expand 2 Items
Protector® Demonstration Hoods, Labconco®
Supplier: Labconco
Designed for classroom use, these lightweight hoods feature an epoxy-coated aluminum frame with tempered safety glass front sash, sides, rear baffle, and top.
Expand 6 Items
Guardian™ Digital Airflow Monitor, Labconco®
Supplier: Labconco
Guardian Digital Airflow Monitors alert the operator to low airflow conditions
Expand 1 Items
REDISHIP Auxiliary Air Chambers, Kewaunee®
Supplier: Kewaunee
These chambers can be added to the top of any Supreme Air fume hood to convert it to an auxiliary air fume hood.
Expand 2 Items
Protector® Stainless Steel Perchloric Acid Laboratory Hoods, Labconco®
Supplier: Labconco
Interior is one piece, Type 316 stainless steel with integral work surface and drainage trough.
Expand 40 Items
Protector® XL™ Floor-Mounted Laboratory Hoods with Horizontal Sashes, Labconco
Supplier: Labconco
Protector XL floor-mounted laboratory hoods with horizontal-sliding sashes are suitable for general chemistry applications and have the dimensions required for over-sized equipment.
Expand 12 Items
Guardian™ Airflow Monitor, Labconco®
Supplier: Labconco
Guardian Airflow Monitors are designed to continuously monitor airflow through Labconco fume hoods