Polyethylene Aspirator Filter Pump
Catalog # 470005-486
Supplier: Bel-Art
Specifications
- Description:Pump, Polyethylene, Aspirator Filter, Scienceware
- Cat. no.:470005-486
- Supplier No.:F329470000
Specifications
About this item
Because fumes and corrosive filter agents do not affect this aspirator, it can be used safely with bases, acids, and strong chemicals. The pump fits any standard laboratory faucet and accepts tubing up to 9.5mm ( 3/8") ID. Operates efficiently on normal water pressure above 11 psi.