Order Entry
Canada
Orders LinkContactUsLinkComponent
Armored Thermometer

Armored Thermometer

Armored Thermometer, 0...+50 °C, 15.5 cm
Catalog # 470101-444
Supplier:   LaMotte
Supplier #:  1066
Armored Thermometer
Catalog # 470101-444
Supplier:  LaMotte
Armored Thermometer
Armored Thermometer

Armored Thermometer, 0...+50 °C, 15.5 cm

Price
57,35 $
Log in for availability
Each
Product Details & Documents

Precision Non-Mercury Thermometer

  • Environmentally safe
  • Engraved graduations on white tubing for increased readability
  • Protective plastic jacket with window opening
Specifications
  • Description:
    Armored Thermometer
  • Length:
    15.5 cm
  • Range:
    0...+50 °C