Aristolochia, Two-Year Stem Slide
470181-332
: Avantor
- Description:Aristolochia, Two-Year Stem
- Cat. no.:470181-332
- Stain:Quadruple
- Supplier No.:WARD470181-332
- Orientation:Cross section
Illustrates Plant Structure
- Identifying features clearly distinguishable
- Rigorous quality control standards
Dutchman’s pipe. Vine. Shows two years' growth of xylem layers (growth rings).