ContactUsLinkComponent
Ancient Microbes, Mixed Types Slide
  470176-986
 :  Avantor
undefined
Ancient Microbes, Mixed Types Slide
  470176-986
 :  Avantor
  • Description:
    Ancient microbes, mixed types
  • Cat. no.:
    470176-986
  • Stain:
    Gram (+/-)
  • Supplier No.:
    WARD470176-986
  • Orientation:
    Smear
Illustrates old type bacteria/Archaea.
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Rods/cocci, involuted forms. Mixture of various types of 'ancient bacteria' representing extreme halophiles (Halobacterium salinarium (rods), Staphylococcus saprophyticus (coccus), and Cellumonas sp. (rods)).