Order Entry
Canada
Orders LinkContactUsLinkComponent
Antimony Trichloride

Antimony Trichloride

Antimony Trichloride, Reagent, 100 g
Catalog # 470300-280
Supplier:   Ward's Science
Supplier #:  AA0376-100GR
Antimony Trichloride
Catalog # 470300-280
Supplier:  Ward's Science
CAS Number:  
Antimony Trichloride
Antimony Trichloride

Antimony Trichloride, Reagent, 100 g

CAS Number:  
Price
$175.95
Log in for availability
Each (100g)
Minimum order quantity is 1
Product Details & Documents

CAS Number: 10025-91-9
Formula Weight: 228.11
Formula: SbCl3
Density (g/mL): 3.1
Boiling Point (°C): 223
Freezing Point (°C): 73.4
Solubility: Alcohol, Acetone, Acids, and Reactive with Water
Synonyms: Antimony(III) Chloride
Shelf Life (months): 12
Storage: White

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Specifications

Specifications

  • Description:
    Antimony Trichloride
  • Size:
    100 g
  • Chemical/physical specifications:
    Crystal
  • Grade:
    Reagent

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.