Angiosperm Anatomy Slide Set
Angiosperm Anatomy Slide Set, 12, Plant
Catalog # 470183-480
Supplier: Avantor
Jump to:
Product Details & Documents
Explore the Anatomy of Flowering Plants
- Identifying features clearly distinguishable
- Rigorous quality control standards
Ordering information: Set includes Monocot & Dicot Roots (cs), Stems (cs), Leaves (cs), Leaf Epidermis (cs) and Flower buds (cs); Allium mitosis (sect); Mixed pollen (wm); Lillium ovary (sect); Zea (corn), kernel (sect); Lillium anther (sect); Capsella (shepherd’s purse), mature embryo (sect); Gymnosperm & angiosperm stem (sect).
Specifications
- Type:Plant
- Description:Angiosperm Anatomy Slide Set
- No. of slides:12