Orders LinkContactUsLinkComponent
Kemtec™ Amino Acid Analysis by TLC Lab Activity.
Kemtec™ Amino Acid Analysis by TLC Lab Activity.
Supplier:   Kemtec
Kemtec™ Amino Acid Analysis by TLC Lab Activity.
Supplier:  Kemtec
Certificates
Kemtec™ Amino Acid Analysis by TLC Lab Activity.
Kemtec™ Amino Acid Analysis by TLC Lab Activity.
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Explore the Theory of Chromatography.
  • Visually determine the unknowns
  • Target grades 10-14
Students learn the theory of chromatography; then separate two different unknown amino acid mixtures using thin layer chromatography (TLC) with amino acid standards run alongside the unknowns. Ninhydrin is used as a visualizing agent; then the rate of flow (Rf) values are calculated for each spot identified on the chromatograms. Students are challenged to predict the migration of the amino acids on the TLC after looking up the structures and pKa values of the four amino acid standards. The unknown components are identified by comparing the Rf values from the unknown amino acid mixtures to Rf data obtained from the four known amino acid standards. Designed for 24 students working in six groups of 4.
Ordering information: Kit includes Instructor Manual, Student Manual, Student Data Sheets, Amino Acid Standards, Unknown Amino Acid Mixture I, Unknown Amino Acid Mixture II, Silica Gel Strips, Chromatography Solvent, Toothpicks, Foil to cover Chromatography Chambers, Ninhydrin, Acetone, Spray Bottles. Hair Dryer, 600 mL beakers for chromatography chambers, clear plastic rulers, and standard laboratory supplies are required but not included.
Delivery information: Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.
Documents
Product Family Options
Product Information
  • Includes
    No. of lab groups
    Availability
    Price
  • 470024-062
    Amino Acid Analysis by TLC Lab Activity
    Amino Acid Analysis by TLC Lab Activity
    Teacher's Guide and Student Copymaster
    6
    Discontinued
    Supplier Number: -
  • 470024-304
    Amino Acid Analysis By TLC Lab Activity, Refill Kit
    Amino Acid Analysis By TLC Lab Activity, Refill Kit
    Log in for availability
    Log in for availability
    Kit of 1
    Kit of 1 - $593.30
    $593.30
    Kit of 1
       $593.30
    Supplier Number: 8-164R
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)