Orders LinkContactUsLinkComponent
Amphioxus Slide
Amphioxus Slide
Amphioxus, Whole mount, Schneider's and Fast Green
Catalog # 470181-686
Supplier:   Avantor
Amphioxus Slide
Catalog # 470181-686
Supplier:  Avantor
Amphioxus Slide
Amphioxus Slide
Amphioxus, Whole mount, Schneider's and Fast Green
Price
$23.60
Log in for availability
Each
Product Details & Documents
Explore early animal development stages
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Small adult exhibiting three main chordate characteristics: notochord, nerve chord, and pharyngeal clefts. It resembles both the echinoderms and vertebrates, providing a unique specimen for embryological study.
Specifications
  • Description:
    Amphioxus
  • Stain:
    Schneider's and Fast Green
  • Orientation:
    Whole mount