Advanced Plant Slide Set
Catalog # 470183-486
Supplier: Avantor
Specifications
- Type:Plant
- Description:Advanced Plant Slide Set
- No. of Slides:100
- Cat. no.:470183-486
- Supplier No.:WARD470183-486
Specifications
About this item
Study Cellular and Tissue Organization of Plant Anatomy and Phylogenetic Comparison
- Identifying features clearly distinguishable
- Rigorous quality control standards
Ligustrum and Nymphaea leaves showing the variation in structure.
Ordering information: Set Includes 100 slides allowing an in-depth exploration of the diverse plant kingdom. Slides include the following types of slides from different species: protective wood fibers, stems; vascular system; root system; reproductive structures; pollen and embryos.