Order Entry
Canada
ContactUsLinkComponent
Advanced pH Meter Kit, Sper Scientific
Advanced pH Meter Kit, Sper Scientific
Catalog # 470204-174
Supplier:  Sper Scientific
undefined
Advanced pH Meter Kit, Sper Scientific
Catalog # 470204-174
Supplier:  Sper Scientific
Supplier Number:  850056

Specifications

  • Temperature range:
    23 - 176 °F; -5 - 80 °C
  • Description:
    Advanced pH Meter Kit
  • Temperature accuracy:
    ±2°F; ±1°C
  • Temperature resolution:
    0.1°F/°C
  • W×D×H:
    2 3/4 × 1 1/4 × 7"
  • mV accuracy:
    ±2 mV; ±0.2 mV
  • mV range:
    -499...+499; -199.9...+199.9
  • mV resolution:
    1 mV
  • pH accuracy:
    ±0.02
  • pH range:
    0 - 14
  • pH resolution:
    0.01
  • Cat. no.:
    470204-174
  • Supplier No.:
    850056

Specifications

About this item

Reads either pH (in 0.01 resolution) or mV in large digits, with time, date, and temperature (in °C or °F) shown simultaneously on the lower display.

  • Measures multiple conditions
  • Durable Housing
  • Easy to Read

Also indicates stable reading, low battery and displays calibration data. Scroll 99 data points with time stamps and min/max readings directly on the backlit LCD. Features hold, up to 5 point calibration, automatic buffer recognition, manual or automatic temperature compensation, and a USB port.

pH Range: 0 - 14. Resolution: 0.01pH. Accuracy: ±0.02pH.
mV Range: -499 to +499mV; -199.9 to +199.9mV. Resolution: 1mV; 0.1mV. Accuracy: ±2mV; ±0.2mV
Temperature Range: 23 - 176°F; -5 - 80°C Resolution: 0.1°F/°C. Accuracy: ±2°F; ±1°C

Ordering information: 5-year warranty. Kit includes Advanced pH Meter model 850055 in a hard carrying case complete with a detachable temperature compensated pH probe with 3½’ cable, pH buffers 4,7,10 and distilled water.