ContactUsLinkComponent
Acrylic Refraction Cube
Acrylic Refraction Cube
Catalog # 470149-494
undefined
Acrylic Refraction Cube
Catalog # 470149-494
Supplier Number:  P30347
Specifications
  • Description:
    Acrylic Refraction Cube
  • Environmentally Preferable:
  • Cat. no.:
    470149-494
  • Supplier No.:
    P30347
Specifications
About this item
Ideal for Index of Refraction and Total Internal Reflection Experiments
The 5cm faces on this acrylic cube are transparent and highly polished for clarity. The cube has an index of refraction of about 1.5.