ContactUsLinkComponent
Acrylic Lens and Prism Set
Acrylic Lens and Prism Set
Catalog # 470110-634
undefined
Acrylic Lens and Prism Set
Catalog # 470110-634
Supplier Number:  OPSETP3
Specifications
  • Description:
    Acrylic Lens and Prism Set, 7 pcs.
  • Cat. no.:
    470110-634
  • Supplier No.:
    OPSETP3
Specifications
About this item
This set is economical, durable, and versatile.
  • Clear Acrylic Elements
  • Variety of Lens and Prisms
  • Economical
  • Housed in a Convenient Plastic Container
  • Durable
Housed in a convenient plastic container, this set includes seven clear acrylic elements.
Each elements has one frosted surface that reveals the path of a light beam passing through.
Ordering information: This kit includes a double-convex lens, double-concave lens, semicircular lens, trapezoidal lens, small right-angle prism, equilateral prism, and large right-angle prism.