Order Entry
Canada
Orders LinkContactUsLinkComponent
Plastic Slide Box for 25 Slides

Plastic Slide Box for 25 Slides

Plastic Slide Box, 25, 5.5 x 3.5 x 1.3"
Catalog # 470156-718
Supplier:   C & A Scientific
Supplier #:  97-0025
Plastic Slide Box for 25 Slides
Catalog # 470156-718
Supplier:  C & A Scientific
Plastic Slide Box for 25 Slides
Plastic Slide Box for 25 Slides

Plastic Slide Box, 25, 5.5 x 3.5 x 1.3"

Price
$12.15
Log in for availability
Each Case of 200
Product Details & Documents

Store your slides efficiently.

  • Store your slides safely and keep them well organized

The plastic box protects 1" x 3" slides and won’t warp when the temperature and humidity vary. It is stackable, with telescoping covers, a cork bottom, and embossed slide numbers on the bottom.

Specifications
  • Capacity:
    25
  • Description:
    Plastic Slide Box
  • Dimensions:
    5.5 x 3.5 x 1.3"
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.