ContactUsLinkComponent
12" Engineer’s Ruler
12" Engineer’s Ruler
Catalog # 470020-158
Supplier:  Acme United
undefined
12" Engineer’s Ruler
Catalog # 470020-158
Supplier:  Acme United
Supplier Number:  M-100
Specifications
  • Description:
    12" Engineer’s ruler
  • Material:
    Plastic
  • Cat. no.:
    470020-158
  • Supplier No.:
    M-100
Specifications
About this item
This clear plastic ruler combines three scales in one.
The outer edges are marked in ¹/₁₀ and ¹/₁₆" graduations, while the center is in millimeters.