ContactUsLinkComponent
Specific Gravity Pycnometer Tube, Ace Glass Incorporated
Specific Gravity Pycnometer Tube, Ace Glass Incorporated
Catalog # 89051-716
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
undefined
Specific Gravity Pycnometer Tube, Ace Glass Incorporated
Catalog # 89051-716
Supplier:  Ace Glass
Supplier Number:  5437-12
Product available on GSA Advantage®
Small Business Enterprise
Specifications
  • Description:
    Specific Gravity Pycnometer Tube
  • Cat. no.:
    89051-716
  • Supplier No.:
    5437-12
Specifications
About this item
This tube is a method for measurement of light hydrocarbon liquids as per ASTM D 941
The capacity of the bulb is 4.5mL. Fabricated from borosilicate glass.