Order Entry
Canada
ContactUsLinkComponent
 
Beaker Mug, Graduated, Ace Glass Incorporated
  89082-898
 :  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
undefined
Beaker Mug, Graduated, Ace Glass Incorporated
  89082-898
 :  Ace Glass
 :  5324-10
Product available on GSA Advantage®
Small Business Enterprise

 

  • Capacity:
    400 ml (14 oz.)
  • Description:
    Heavy-Wall Graduated Beaker Mug
  • Cat. no.:
    89082-898
  • Supplier No.:
    5324-10

 

 

This is a 400ml heavy-wall, graduated beaker mug

Fabricated from borosilicate glass.