Order Entry
Canada
ContactUsLinkComponent
GlycoLink™, Pierce™ Immobilization Kits and Resin, Thermo Scientific
GlycoLink™, Pierce™ Immobilization Kits and Resin, Thermo Scientific
Catalog # CAPI88942
Supplier:  Thermo Scientific
CAS Number:  
GlycoLink™, Pierce™ Immobilization Kits and Resin, Thermo Scientific
Catalog # CAPI88942
Supplier:  Thermo Scientific
Supplier Number:  88942
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    GlycoLink Micro Immobilization Kit
  • Size:
    10-column kit
  • Cat. no.:
    CAPI88942
  • Supplier No.:
    88942
  • Sufficient For:
    Immobilizing 50 to 500 µg of glycoprotein per column

Specifications

About this item

Hydrazide-activated resin to immobilize glycoproteins for affinity purification.

  • High-capacity—immobilize 1 to 10mg of oxidized antibody or other glycoprotein per milliliter of resin
  • Fast and efficient—immobilize in as few as 30 minutes; achieve at least 90% coupling of most glycoproteins in less than 4 hours (efficiency depends on the amount and type of glycosylation)
  • Specific immobilization—the hydrazide-activated UltraLink Resin conjugates only to purified glycoproteins whose sugar (e.g., sialic acid) groups have been gently oxidized with periodate
  • Stable attachment—resonance structure of the hydrazone bonds are sufficiently stable for multiple affinity purifications with one batch of prepared resin; no stabilization step is required
  • Preserves antibody function—immobilizes IgG via the Fc region, thereby keeping both antigen binding sites available for capturing target
  • Convenient sizes—choose from two complete 10-column kits: one with 1mL columns and one with 0.1mL columns

Thermo Scientific GlycoLink Immobilization Kits provide affinity columns and reagents to immobilize glycoproteins through oxidized sugar groups for use in affinity-purification applications or studies of protein-protein interactions.

The GlycoLink Kits use hydrazide-activated resin and a special catalyst to prepare affinity columns with polyclonal antibodies, which typically have abundant carbohydrates (glycosylation) on their Fc portions. Any glycosylated proteins (even monoclonal antibodies) that contain periodate-oxidizable carbohydrate groups can be immobilized with these kits to make reusable affinity columns. Antibodies immobilized by this hydrazide immobilization chemistry have unobstructed antigen-binding sites and optimal purification capability. Prepared columns with stable glycoproteins or antibodies can be regenerated and reused at least five times for affinity-purification without significant loss in binding capacity.

Kit Content for PI88941:
GlycoLink Columns, 1mL, 10 columns
GlycoLink Coupling Buffer, 2 x 100mL
Aniline, 0.2mL
Wash Buffer, 120mL
Sodium meta-periodate, 0.5g
Zeba Spin Desalting Columns, 5mL, 10 columns
PBS Pack (makes 500mL), 1 pack
Column Accessories, 10 ea
Kit Content for PI88942:
GlycoLink Columns, 0.1mL, 10 columns
GlycoLink Coupling Buffer, 100mL
Aniline, 0.2mL
Wash Buffer, 60mL
Sodium meta-periodate, 0.5g
Zeba Spin Desalting Columns, 0.5mL, 10 columns
PBS Pack (makes 500mL), 1 pack
Column Accessories, 10 ea
Kit Content is N/A for PI53149.