Orders LinkContactUsLinkComponent
NMC Valve Wheel Lockouts
NMC Valve Wheel Lockouts
NMC Valve Wheel Lockouts
Certificates
NMC Valve Wheel Lockouts
NMC Valve Wheel Lockouts
5 Options Available Below
Product Details & Documents
Lockouts consist of two flattened half moons that completely cover the valve wheels from 1" to 14" in diameter
Made from strong Polypro plastic, fully dielectric. Highly resistant to cracking and abrasion. Resistant to extreme temperature changes 50⁰F
Product Family Options
Product Information
  • Availability
    Price
  • 94022-904
    Fits 2.54 cm (1") To 6.35 cm (2.5") Dia.
    Fits 2.54 cm (1") To 6.35 cm (2.5") Dia.
    Log in for availability
    Log in for availability
    Each
    Each - $98.92
    $98.92
    Each
       $98.92
    Minimum order quantity is 1
    Supplier Number: VL02R
  • 94022-906
    Fits 6.35 cm (2.5") To 12.7 cm (5") Dia.
    Fits 6.35 cm (2.5") To 12.7 cm (5") Dia.
    Log in for availability
    Log in for availability
    Each
    Each - $163.85
    $163.85
    Each
       $163.85
    Minimum order quantity is 1
    Supplier Number: VL04R
  • 94022-908
    Fits 12.7 cm (5") To 16.51 cm (6.5") Dia.
    Fits 12.7 cm (5") To 16.51 cm (6.5") Dia.
    Log in for availability
    Log in for availability
    Each
    Each - $199.34
    $199.34
    Each
       $199.34
    Minimum order quantity is 1
    Supplier Number: VL06R
  • 94022-910
    Fits 16.51 cm (6.5") To 25.4 cm (10") Dia.
    Fits 16.51 cm (6.5") To 25.4 cm (10") Dia.
    Log in for availability
    Log in for availability
    Each
    Each - $248.90
    $248.90
    Each
       $248.90
    Minimum order quantity is 1
    Supplier Number: VL09R
  • 94022-912
    Fits 25.4 cm (10") To 35.56 cm (14") Dia.
    Fits 25.4 cm (10") To 35.56 cm (14") Dia.
    Log in for availability
    Log in for availability
    Each
    Each - $481.30
    $481.30
    Each
       $481.30
    Minimum order quantity is 1
    Supplier Number: VL13R