Master Lock® Circuit Breaker Switch Padlock, NMC
Catalog # 94017-522
Supplier: ACCUFORM MANUFACTURING, INC
Specifications
- Description:3.35cm (1⅛") Steel Body W/ 12.7cm (5") Cable
- Environmentally Preferable:
- Cat. no.:94017-522
- Supplier No.:MLC
Specifications
About this item
Provides the flexibility to effectively lockout breaker switches
Flexible steel cable allows padlock to lock all types of circuit breaker lockout devices. Flex-cable allows the lock body to be moved out of the way and panel door to be closed properly.