Master Lock® Adjustable Cable Lockout, NMC
.64cm (.25") steel cable with vinyl coating
Catalog # 94007-602
Supplier: ACCUFORM MANUFACTURING, INC
Supplier #: CABLO
Master Lock® Adjustable Cable Lockout, NMC
Catalog # 94007-602
Supplier: ACCUFORM MANUFACTURING, INC
Jump to:
Product Details & Documents
Infinitely adjustable for a secure fit every time
Feed the cable end through the points to be locked out, then back through the lockout body. Tough, multi-strand .64cm (¼") dia. Steel cable with clear vinyl coating. Up to 4 workers can apply their locks. Includes a 15.24cm (6") cable.
Specifications
- Description:.64cm (.25") steel cable with vinyl coating