GlobalMark® Reflective Tape, B-584, Brady Worldwide®
Supplier: Brady Worldwide
Certificates
About this item
GlobalMark tapes are easy to install and very durable
Not recommended for plotter cutting. Brady B-584 is a reflective tape designed for use with the Labelizer plus industrial labeling system, GraphicsPro label and sign creator, VersaPrinter label and sign maker system, and the HandiMark portable label maker, PowerMark sign and label maker and GlobalMark industrial label maker. Minimum application temperature: silver: 50°F (10°C), yellow and orange: 0°F (-18°C). Average outdoor durability: 4-6 years (Average expected outdoor life of product will depend on user definition of failure, climatic conditions and mounting techniques.)
Frequently Bought Together