GlobalMark® Transfer Tape, Brady Worldwide®
CA89007-748
- Color:Clear
- Description:Transfer tape
- Material:Transfer Tape
- Size:4.25" W x 300' L
- Cat. no.:CA89007-748
- For Use With:GlobalMark Printer
- Supplier No.:76737
Leaves cut characters pre-spaced for transferring from liner to applying them to desired surface.