GlobalMark® Tape, B-565, Brady Worldwide®
Supplier: Brady Worldwide
Certificates
About this item
GlobalMark tapes are easy to install and very durable
Not supported for plotter cutting. Brady-565 is a metallized polyester film with a permanent acrylic pressure sensitive adhesive and a topcoat specifically formulated for thermal transfer printing and is designed for use in cartridge form with the GlobalMark printer and the BBP31Sign and Label Printer. Matte Metallized Polyester with a permanent pressure sensitive acrylic adhesive. Minimum Application Temperature: 0°F (-18°C).
Customers Who Bought This Also Bought