GlobalMark® Tape, B-581, Brady Worldwide®
Supplier: Brady Worldwide
Certificates
About this item
GlobalMark tapes are easy to install and very durable
Brady B-581 indoor/outdoor marking film is a high gloss, durable, flexible, pressure sensitive adhesive product, designed for use in cartridge form with the Labelizer Plus printer, VersaPrinter Label and Sign Maker System, and the HandiMark Portable Label Maker, and the GlobalMark Industrial Label Maker. Minimum Application Temperature: 50°F (10°C).
Frequently Bought Together