GlobalMark® Cold Temperature Tape, B-549, Brady Worldwide®
Supplier: Brady Worldwide
Certificates
About this item
GlobalMark tapes are easy to install and very durable
Not recommended for plotter cutting. Brady B-549 is a flexible, dimensionally stable, low temperature, pressure sensitive adhesive tape. Brady B-549 is designed for use with the Brady Labelizer Plus Industrial Labeling System and the VersaPrinter Label and Sign Maker System and GlobalMark Industrial Label Maker for permanent application.
Customers Who Bought This Also Bought