GlobalMark® Tape, B-588, Brady Worldwide®
Supplier: Brady Worldwide
Certificates
About this item
GlobalMark tapes are easy to install and very durable
Brady B-588 is a vinyl film with pressure sensitive adhesive designed for use with the GlobalMark Label and MiniMark Sign Printers. Brady B-588 is used for general labeling and marking, indoors and outdoors. Minimum Application Temperature: 50°F (10°C). Service Temperature: -40°F to 176°F (-40°C to 80°C) Average Outdoor Durability: 3-5 years.
Frequently Bought Together