MicroPoint, extended length, ART 10 REACH, Thermo Scientific
Supplier: Thermo Scientific
Certificates
About this item
Molecular BioProducts ART (Aerosol Resistant Tip) specialty pipette tips seal when exposed to potential contaminants, trapping them inside the barrier, providing accurate and consistent performance.
These tips come with a self-sealing filter tip that protects pipettes from crossover contamination by not allowing liquids or aerosols to touch pipettes.
Specifications
- Catalog No:
- 72830-420
- Environmentally Preferable:
- Low Manufacturing Impact
- Environmental certification:
- ISO14001 (Certificate Registration No. EMS 752603)