Dewar Condenser, Angled Top Joint, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
This Dewar condenser is for use with dry ice and other solid cooling agents
It can also be adapted for use as a small constant temperature bath by using a constant boiling liquid. In this manner, it can also be used as an oil bath. Has standard taper 14/20 outer joint on the side at a 105° angle, and standard taper 14/20 inner joint at bottom. Fabricated from borosilicate glass.
Frequently Bought Together