Orders LinkContactUsLinkComponent
Condenser, Allihn
Condenser, Allihn
Supplier:   Ace Glass
Condenser, Allihn
Supplier:  Ace Glass
Certificates
Condenser, Allihn
Condenser, Allihn
12 Options Available Below
Product Details & Documents
Bulb type Allihn condenser, approximately one bulb per every 50 mm, with standard taper inner joint at bottom and reinforced standard taper outer joint at top
Fabricated from borosilicate glass. Size D glass hose connections, for use with 3/8-inch I.D. tubing.
Product Family Options
Product Information
  • [ST] joint
    Jacket length
    Availability
    Price
  • 89053-780
    Condenser, Allihn
    Condenser, Allihn
    24/40
    200 mm
    Log in for availability
    Log in for availability
    Each
    Each - 401,71 $
    401,71 $
    New Customer Promotional Price
    Each
       401,71 $
    Supplier Number: 5945-12
  • 89053-786
    Condenser, Allihn
    Condenser, Allihn
    19/38
    250 mm
    Log in for availability
    Log in for availability
    Each
    Each - 408,22 $
    408,22 $
    New Customer Promotional Price
    Each
       408,22 $
    Supplier Number: 5945-05
  • 89053-794
    Condenser, Allihn
    Condenser, Allihn
    24/40
    400 mm
    Log in for availability
    Log in for availability
    Each
    Each - 498,11 $
    498,11 $
    New Customer Promotional Price
    Each
       498,11 $
    Supplier Number: 5945-15
  • 89053-796
    Condenser, Allihn
    Condenser, Allihn
    24/40
    500 mm
    Log in for availability
    Log in for availability
    Each
    Each - 524,40 $
    524,40 $
    New Customer Promotional Price
    Each
       524,40 $
    Supplier Number: 5945-16
  • 89053-798
    Condenser, Allihn
    Condenser, Allihn
    24/40
    600 mm
    Log in for availability
    Log in for availability
    Each
    Each - 532,18 $
    532,18 $
    New Customer Promotional Price
    Each
       532,18 $
    Supplier Number: 5945-17
  • 89053-800
    Condenser, Allihn
    Condenser, Allihn
    29/42
    300 mm
    Log in for availability
    Log in for availability
    Each
    Each - 505,12 $
    505,12 $
    New Customer Promotional Price
    Each
       505,12 $
    Supplier Number: 5945-24
  • 89053-802
    Condenser, Allihn
    Condenser, Allihn
    29/42
    400 mm
    Log in for availability
    Log in for availability
    Each
    Each - 518,03 $
    518,03 $
    New Customer Promotional Price
    Each
       518,03 $
    Supplier Number: 5945-25
  • 89053-804
    Condenser, Allihn
    Condenser, Allihn
    29/42
    500 mm
    Log in for availability
    Log in for availability
    Each
    Each - 553,20 $
    553,20 $
    New Customer Promotional Price
    Each
       553,20 $
    Supplier Number: 5945-26
  • 89053-806
    Condenser, Allihn
    Condenser, Allihn
    29/42
    600 mm
    Log in for availability
    Log in for availability
    Each
    Each - 636,49 $
    636,49 $
    New Customer Promotional Price
    Each
       636,49 $
    Supplier Number: 5945-27
  • 89053-808
    Condenser, Allihn
    Condenser, Allihn
    24/25
    200 mm
    Log in for availability
    Log in for availability
    Each
    Each - 410,81 $
    410,81 $
    New Customer Promotional Price
    Each
       410,81 $
    Supplier Number: 5945-34
  • 89053-866
    Condenser, Allihn
    Condenser, Allihn
    24/40
    300 mm
    Log in for availability
    Log in for availability
    Each
    Each - 450,86 $
    450,86 $
    New Customer Promotional Price
    Each
       450,86 $
    Supplier Number: 5945-14
  • 89083-320
    Condenser, Allihn
    Condenser, Allihn
    24/40
    250 mm
    Log in for availability
    Log in for availability
    Each
    Each - 423,44 $
    423,44 $
    New Customer Promotional Price
    Each
       423,44 $
    Supplier Number: 5945-13