Order Entry
Canada
ContactUsLinkComponent
Biotin Resin for Isolation of Avidin, Streptavidin & Neutravidin Coupled Molecules, G-Biosciences
Biotin Resin for Isolation of Avidin, Streptavidin & Neutravidin Coupled Molecules, G-Biosciences
Catalog # CA71003-226
Supplier:  G-Biosciences
CAS Number:  
Biotin Resin for Isolation of Avidin, Streptavidin & Neutravidin Coupled Molecules, G-Biosciences
Catalog # CA71003-226
Supplier:  G-Biosciences
Supplier Number:  786-598
CAS Number:  

Specifications

  • Description:
    Immobilized Biotin Resin
  • Size:
    5 mL resin
  • Cat. no.:
    CA71003-226
  • Supplier No.:
    786-598

Specifications

About this item

G-Biosciences' Immobilized Biotin Resin is designed for the high affinity chromatography purifications of avidin, streptavidin and Neutravidin protein.

G-Biosciences' Immobilized Biotin Resin consists of biotin coupled to 6% cross-linked agarose. Biotin, a 244 Dalton vitamin (Vitamin H) molecule, exhibits an extraordinary binding affinity for avidin (Ka=10¹⁵M¯¹) and streptavidin. Biotin and avidin interaction is rapid, and once the bond is established, it can survive up to 3 M guanidine-hydrochloride and extremes of pH. Biotin-avidin bonds can only be reversed by denaturing the avidin protein molecule with 8 M guanidine-hydrochloride at pH 1.5 or by boiling in SDS PAGE sample loading buffer.

G-Biosciences' Immobilized Biotin Resin is covalently coupled to limit leaching, and it is reusable at least 10 times.

Ordering information: Accessories available.