Order Entry
Canada
Orders LinkContactUsLinkComponent
Protein Solubilization Buffer, G-Biosciences

Protein Solubilization Buffer, G-Biosciences

FOCUS™ Protein Solubilization Buffer (PSB) with Diluent III, 50 mL
Catalog # CA95043-574
Supplier:   G-Biosciences
Supplier #:  786-PSB
Protein Solubilization Buffer, G-Biosciences
Catalog # CA95043-574
Supplier:  G-Biosciences
CAS Number:  
Protein Solubilization Buffer, G-Biosciences

FOCUS™ Protein Solubilization Buffer (PSB) with Diluent III, 50 mL

CAS Number:  
Price
$643.63
Log in for availability
Each (50ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

G-Biosciences' FOCUS™ Protein Solubilization Buffer is a dry, urea-based and ready-to-use buffer for protein solubilization

G-Biosciences' FPS Buffer is a DRY, pre-mixed formulation of urea, thiourea, CHAPS and Non-Detergent Sulfobetaine (ND SB) 201 for maximum solubilizing strength. The dry format allows researcher’s to freshly rehydrate as much or as little as is required and therefore prevent urea induced carbamylation.

It is a proprietary modification of urea based chaotropic extraction buffers. Normal urea based buffers solubilize proteins effectively however can modify the native charge of the proteins, due to carbamylation, a process that modifies amino groups, inducing changes in the isoelectric point of proteins leading to artifactual 2D results.

Ordering information: Accessories available.

Specifications

Specifications

  • Description:
    FOCUS™ Protein Solubilization Buffer (PSB) with Diluent III
  • Size:
    50 mL

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.