You searched for: Petri Dishes
Standard Petri dishes have bubble-free construction to evenly spread cell growth. High-quality plates have a translucent longevity for high-resolution microscopic studies Disposable or reusable petri dish surfaces come non-treated or treated for cell attachment. Multi-well designs allow simple side-by-side comparisons. Graduated models easily locate specimen even in large culture media colonies. Tight fitting lids protect sample from contamination and may be vented. Aspetic, sterile, and non-sterile Petri dishes come in any growth area size needed.
Slippable Petri Dishes, 100 mm
Supplier: VWR International
These slippable polystyrene Petri dishes provide optimal flatness.
Expand 2 Items
VWR® Petri Dishes
Supplier: VWR International
Our high-quality borosilicate Petri dishes meet ASTM standards and are a great value.
Expand 2 Items
VWR® Disposable Petri Dishes
Supplier: VWR International
Clearly superior, high-quality Petri dishes are manufactured from virgin crystal grade polystyrene for optimum flatness and high optical clarity.
Expand 37 Items
Petri Dishes
Supplier: VWR International
An assortment of VWR® petri dishes for laboratory applications are available.
Expand 5 Items
VWR® Petri Dishes, Glass
Supplier: VWR International
These Petri dishes are suitable for use in pharmaceuticals, food processing, hospital environment and other industries for samples purposes.
Expand 9 Items
VWR® Petri Dishes with Markings
Supplier: VWR International
These Petri dishes are ideal for antibiotic susceptibility testing.
Expand 1 Items
VWR® Conventional Petri Dishes
Supplier: VWR International
These dishes offer distortion-free viewing, making it ideal for microscopic examination.
Expand 1 Items
VWR® Deep Petri Dishes
Supplier: VWR International
These deep-dish Petri dishes accommodate deeper media fills for longer culturing periods.
Expand 1 Items
VWR® High Profile Petri Dishes
Supplier: VWR International
These high-profile Petri dishes are specifically designed to accommodate more media.
Expand 1 Items
VWR® Petri Dishes for Single Membrane Filters
Supplier: VWR International
These dishes save space in refrigerators and incubators.
Expand 2 Items
VWR® Petri Dishes, Nesting Alpine
Supplier: VWR International
The unique recessed lid on these dishes offers excellent stacking stability and saves space.
Expand 1 Items
VWR® Petri Dish Sets (Tops and Bottoms)
Supplier: VWR International
Our high-quality borosilicate Petri dishes meet ASTM standards and are a great value.
Expand 2 Items
VWR® Sectional Petri Dishes
Supplier: VWR International
These sectional Petri dishes are ideal for simultaneous differential studies of organisms or media in a common environment.
Expand 1 Items
VWR® Conventional Petri Dishes, Semi Stacking
Supplier: VWR International
These semi-stacking Petri dishes are available with a semi-stacking ring for easy transit during automatic filling or in a sliding version with a totally smooth lid.
Expand 2 Items
Budget Antibiotic Disks
Supplier: Ward's Science
Users can observe the sensitivity of various bacteria and fungi to an array of antibiotics with these convenient kits.
Expand 10 Items
Petri Dishes, Sterile, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
PS dish, cellulose pad. Ideal for microbiological analysis when performing the membrane filter technique. Petri dishes with absorbent pads can be used with broth media, or users can pour agar into dishes without absorbent pads. Accepts 47 mm membrane filter.
Expand 3 Items
Analyslide® Petri Dish, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
The Analyslide® Petri dish has a rectangular base and a circular chamber that accepts a 47 mm membrane filter. The sample is protected during microscopic examination, and membrane filters can be stored and protected for reference.
Expand 1 Items
Corning® Gosselin™ Petri Dish, Corning
Supplier: Corning
Transparent PS Petri dishes for food industry laboratories.
Expand 1 Items
Nalgene® Transparent Petri Dishes, Thermo Scientific
Supplier: Thermo Fisher Scientific
Polymethylpentene for superior chemical and impact resistance
Expand 1 Items
Parter Medical Products Petri Dish, Hardy Diagnostics
Supplier: Hardy Diagnostics
The high quality of Parter Medical's Petri dishes is the result of dedication to product excellence, innovative product design and consumer satisfaction.
Expand 1 Items
Corning® Gosselin™ Petri Dishes, Corning
Supplier: Corning
Contact plates used in hygiene control within the pharmaceutical or food processing industry, as well as in hospital environments. Manufactured in a Class 100 environment with cleanroom packaging. Available aseptic or sterile (SAL 10⁻⁶).
Expand 8 Items
Parter Medical Products Petri Dish, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
Optically clear polystyrene - for distortion free viewing.
Expand 1 Items
Falcon® Petri Dishes, 50×9 mm, Tight-Fit Lid, Sterile, Corning
Supplier: Corning
PS. Manufactured in accordance with the current FDA Quality System.
Expand 1 Items
Nunc® Lab-Tek™ Extra-Depth Disposable Petri Dishes, Thermo Scientific
Supplier: Thermo Fisher Scientific
Culture fungi, bacteria and other microorganisms withThermo Scientific™ Nunc™ Lab-Tek™ Extra-Depth Disposable Petri Dishes.
Expand 1 Items
Falcon® Petri Dishes, Tight-Fit Lid, Sterile
Supplier: Corning
Dishes are constructed with crystal-grade polystyrene and a flat bottom surface for distortion-free optics
Expand 4 Items
Petri dishes, soda lime silca glass, Corning®
Supplier: Corning
Flat, clear petri dishes are manufactured from soda lime silica glass and will withstand repeated sterilization (wet or dry)
Expand 5 Items
Kimax® Petri Dish Sets, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Cell culture dishes feature reinforced beaded edges to resist mechanical breakage and help center the dish cover
Expand 3 Items
Nunc® IVF Centerwell Dish, Thermo Scientific
Supplier: Thermo Fisher Scientific
Thermo Scientific Nunc® provide an extensive portfolio of products intended for use in human In Vitro Fertilization (IVF)
Expand 1 Items
Petri Dish, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
Petri dish, with smooth bottom, without stacking ring.