Order Entry
Canada
ContactUsLinkComponent
Ethidium bromide 0.625 mg/mL in aqueous solution
Ethidium bromide 0.625 mg/mL in aqueous solution
Catalog # CA89168-116
Supplier:  G-Biosciences
CAS Number:  
Ethidium bromide 0.625 mg/mL in aqueous solution
Catalog # CA89168-116
Supplier:  G-Biosciences
Supplier Number:  R041
CAS Number:  

Specifications

  • Size:
    5 mL
  • Cat. no.:
    CA89168-116
  • Supplier No.:
    R041

Specifications

About this item

 :
 :394.31 g/mol
CAS Number:1239-45-8
Formula:C₂₁H₂₀BrN₃
MDL Number:MFCD00011724