Orders LinkContactUsLinkComponent
SpectraDrop™ Plate Systems, Molecular Devices
SpectraDrop™ Plate Systems, Molecular Devices
Supplier:  
Certificates
SpectraDrop™ Plate Systems, Molecular Devices
SpectraDrop™ Plate Systems, Molecular Devices
0 Options Available Below
Product Details & Documents
Molecular Devices’ unique SpectraDrop™ Micro-Volume Plate offers a high throughput and accurate solution for low volume measurement.
  • High throughput
  • Innovative design
  • Accelerated sample prep
  • Increased productivity
Innovative and flexible design features enable accelerated sample preparation time and increased laboratory productivity of DNA, RNA, and protein samples as low as 2 μL
The SpectraDrop™ Micro-Volume Plate incorporates a specially designed adapter and slide pair providing uniform multi-sample assembly. The two-slide design makes the cleaning process a easy and reliable. The users can quickly wipe each slide, autoclave them, sonicate or simply replace them without having to worry about any calibration steps. The simplicity of this arrangement also allows each user to have their own slide combination, making it ideal for multi-user laboratories.
The SpectraDrop™ Micro-Volume Plate comes in two density configurations, 24-wells per plate and 64-wells per plate, and two volume options, 2 or 4 μL, allowing users to select the configuration ideal for their sensitivity, volume and throughput needs. The SpectraDrop™ design never requires any calibration.
The SpectraDrop™ Micro-Volume Plate assures uniform and reproducible analysis and integrates seamlessly with all UV capable SpectraMax Readers - giving you the cost effective option of adding low volume detection with the high standard of data performance of Molecular Devices' readers.
Product Family Options
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • SpectraDrop™ Micro-Volume Microplate 24-Well Kit
    SpectraDrop™ Micro-Volume Microplate 24-Well Kit
    89212-410
    Log in for availability
    Each - $9,248.26
Product Information
  • Catalog #
    Availability
    Price
  • SpectraDrop™ Micro-Volume Microplate HTS Kit
    SpectraDrop™ Micro-Volume Microplate HTS Kit
    89212-412
    Log in for availability
    Each - $14,541.09
Product Information
  • Catalog #
    Availability
    Price
  • SpectraDrop™ Low-Volume Slide 24-Well Refill Kit
    SpectraDrop™ Low-Volume Slide 24-Well Refill Kit
    CA89212-414
    Log in for availability
    Pack of 3 - $822.21
Product Information
  • Catalog #
    Availability
    Price
  • SpectraDrop™ Low-Volume Slide 64-Well Refill Kit
    SpectraDrop™ Low-Volume Slide 64-Well Refill Kit
    CA89212-416
    Log in for availability
    Pack of 3 - $1,986.43
Product Information
  • Catalog #
    Availability
    Price
  • SpectraDrop™ 0.5 mm Pathlength Cover Slide Refill Kit
    SpectraDrop™ 0.5 mm Pathlength Cover Slide Refill Kit
    CA89212-418
    Log in for availability
    Pack of 3 - $822.21
Product Information
  • Catalog #
    Availability
    Price
  • SpectraDrop™ 1.0 mm Pathlength Cover Slide Refill Kit
    SpectraDrop™ 1.0 mm Pathlength Cover Slide Refill Kit
    CA89212-420
    Log in for availability
    Pack of 3 - $822.21