Nalgene Magnetic Carboy Stirrer, Thermo Scientific
Catalog # 66186-041
Supplier: Thermo Fisher Scientific
Specifications
- Environmentally Preferable:
- Cat. no.:66186-041
- Supplier No.:DS2227-0020
- Maximum Stir Bar Length:75 mm
Specifications
About this item
Nalgene Magnetic Carboy Stirrer is used with Nalgene 10- and 20L carboys for low-speed mixing of high volumes of media and buffer solution
The stirrer adjusts easily to fit either a 10 or 20L Nalgene carboy with an 83B size closure. The magnetic stirrer package includes closure and two impellers. Stir bar is designed as a 13mm diameter and shaft O.D: 13mm.