Order Entry
Canada
ContactUsLinkComponent
Modern Round Bottles, High-Density Polyethylene, Narrow Mouth, Qorpak®
Modern Round Bottles, High-Density Polyethylene, Narrow Mouth, Qorpak®
Catalog # 89204-898
Supplier:  Qorpak
Small Business Enterprise
undefined
Modern Round Bottles, High-Density Polyethylene, Narrow Mouth, Qorpak®
Catalog # 89204-898
Supplier:  Qorpak
Supplier Number:  PLA-03260
Small Business Enterprise

Specifications

  • Cap Size:
    28-410
  • Capacity:
    473 ml (16 oz.)
  • Color:
    Natural
  • Material:
    HDPE
  • Cat. no.:
    89204-898
  • Supplier No.:
    PLA-03260

Specifications

About this item

Natural HDPE modern round bottles feature side panels to allow for label application.

The sloped shoulder of this bottle allow for maximum fill capacity.