Order Entry
Canada
ContactUsLinkComponent
Bottles, Oval, Graduated, with Cap, Clear
Bottles, Oval, Graduated, with Cap, Clear
Catalog # 89096-622
Supplier:  Qorpak
Small Business Enterprise
undefined
Bottles, Oval, Graduated, with Cap, Clear
Catalog # 89096-622
Supplier:  Qorpak
Supplier Number:  GLC-13070
Small Business Enterprise

Specifications

  • Cap Size:
    24-400
  • Capacity:
    177 ml (6 oz.)
  • Color:
    Clear
  • Description:
    Black Phenolic with Pulp/Vinyl Liner
  • Material:
    Glass
  • Cat. no.:
    89096-622
  • Supplier No.:
    GLC-13070

Specifications

About this item

Clear oval prescription bottles are graduated in milliliters and ounces with attached black phenolic cap and pulp/vinyl liner.