À propos de cet article
G-Biosciences' GET™ CLEAN DNA kit uses high binding affinity GET™ Spin Columns to remove exess salts, enzymes, unincorporated nucleotides, radiolabels, and primer-dimers from any DNA preparation of 100 base pairs to >20kb
G-Biosciences' GET™ Spin Columns have an enhanced binding affinity for DNA, thus eliminating loss or damage of DNA. The DNA cleaning protocol takes as little as 5 minutes and yields high DNA recovery. No toxic phenol/chloroform extractions or alcohol precipitations are needed.
G-Biosciences' GET™ CLEAN DNA kit is available in a 50 or 100 prep size.
Ideal applications include: PCR clean up, purifying DNA fragments after in-vitro labeling reactions, removal of restriction enzymes from plasmid DNA prior to in-vitro transcription, and cleaning any DNA preparations from interfering agents.
Spécifications
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Amplified/modified DNA
- Sample type:PCR and other enzymatic reactions
- Target molecule:DNA
- Description:GET™ CLEAN DNA