Order Entry
Canada
ContactUsLinkComponent
 
OmniTemplate™ for Single Tube Preparation of PCR Template DNA, G-Biosciences
Catalogue # CA82023-256
Fournisseur:  G-Biosciences
Numéro CAS:  
undefined
OmniTemplate™ for Single Tube Preparation of PCR Template DNA, G-Biosciences
Catalogue # CA82023-256
Fournisseur:  G-Biosciences
Supplier Number:  786-013
Numéro CAS:  

Spécifications

  • Method / Format:
    Extraction for direct PCR
  • Prep size:
    Scalable
  • Refine target molecule:
    Total DNA
  • Sample type:
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    DNA
  • Description:
    OmniTemplate™
  • Size:
    100 Preps
  • Description:
    OmniTemplate™
  • Cat. no.:
    CA82023-256
  • Supplier No.:
    786-013

Spécifications

À propos de cet article

G-Biosciences' OmniTemplate™ is specifically designed for the rapid isolation of a DNA template for polymerase chain reaction (PCR) analysis from mammalian tissue samples, blood and cell cultures

G-Biosciences' OmniTemplate™ method involves isolation of nuclei and their subsequent digestion with LongLife™ Proteinase K in the same tube the samples were collected in. The isolated nuclei are digested to release template DNA in a buffer compatible with PCR and other downstream applications. The OmniTemplate™ samples can be directly used in PCR and other applications without further manipulations. One prep is approximately 1-10 mg tissue.

This single tube method produces a high concentration, ready-to-use supply of DNA template for microanalysis, genotyping and a multitude of other applications.

Ordering information: Accessories available.