About this item
G-Biosciences' GET™ DNA Template is a spin column format kit suitable for the preparation of DNA templates from blood, cells, fungal and animal tissue samples
G-Biosciences' GET™ DNA Template purifies mammalian genomic DNA free from impurities known to inhibit downstream applications. The high yield, extracted DNA is normally >50kb in size. The yield is ~1-40 µg DNA per preparation, depending on the source and quantity used. The kit is supplied as a micro kit for 50 preps and the large kit for 100 preps. The isolated genomic DNA is suitable for PCR and other applications.
G-Biosciences' GET™ Plant DNA Template is a spin column format kit suitable for the preparation of DNA templates from plant samples. The method involves solubilization in Template Extraction Buffer and rapid purification of genomic DNA with a specific genomic DNA binding spin column.
G-Biosciences' GET™ Plant DNA Template purifies plant genomic DNA free from plant impurities known to inhibit downstream applications. The high yield, extracted DNA is normally >50kb in size. The yield is ~30 µg DNA per preparation, depending on the plant source and quantity used. The kit is suitable for 100 preps of 50-100 mg plant tissue. The isolated genomic DNA is suitable for PCR and other applications.
The method involves solubilization in Template Extraction Buffer and rapid purification of genomic DNA with a specific genomic DNA binding spin column.
Specifications
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Total DNA
- Sample type:Plant samples
- Target molecule:DNA
- Description:GET™ Plant DNA Template