femtoPBST™ Wash Buffer, G-Biosciences
Catalog # CA82022-574
Supplier: G-Biosciences
CAS Number:
Specifications
- Description:femto PBST™ [10X]
- Size:250 mL
- Cat. no.:CA82022-574
- Application:Washing Buffer to Replace Standard PBST Buffers
- Supplier No.:786-162
Specifications
About this item
G-Biosciences' femto PBST™ will enhance sensitivity of your immunoassays by minimizing the "washing out" of immuno agents or immuno complexes, a common problem associated with classical PBST buffers
G-Biosciences' femto PBST™ can be used for all your incubation and washing procedures. It is supplied as a 10X solution.
The wash buffers aid in the generation of cleaner backgrounds, resulting in a larger ratio of signal to background.
Ordering information: Accessories available.