Nalgene® Diagnostic Bottles, Natural PPCO, without Closures, Bulk pack, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Nalgene natural PPCO diagnostic bottles are used in test kits and many small-volume applications.
The diagnostic bottles are non-sterile, and are made of polypropylene copolymer. They can be used for sample storage and shipping of small volume, valuable reagents. Closures sold separately.
Ordering information: Bulk Packed for Packaging Applications. For polypropylene caps, see 16060-026 series.
Specifications
- Catalog No:
- 16060-030
- 16060-032
- 16060-034
- Environmentally Preferable:
- Low Manufacturing Impact
- Environmental certification:
- ISO14001 (Certificate Registration No. EMS 752603)