Nalgene® Diagnostic Bottles, Opaque Amber, HDPE, without Closures, Bulk Pack, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Nalgene opaque amber high density polyethylene diagnostic bottles are used in test kits and many small-volume applications.
They have durable uniform walls that are resistant to splitting or puncturing. Diagnostic bottles can be used for sample storage, and shipping of valuable reagents that are light sensitive. Bulk pack comes without closures.
Ordering information: Bulk packed for packaging applications. For polypropylene caps, see 16060-026 series.
Specifications
- Catalog No:
- 16060-044
- 16060-040
- 16060-042
- Environmentally Preferable:
- Low Manufacturing Impact
- Environmental certification:
- ISO14001 (Certificate Registration No. EMS 752603)