Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Acetone 99+%

Flammable
Irritant
   Sustainable Options Available
Acetone 99+%
   Sustainable Options Available
Certificates
Acetone 99+%

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Purity: 99+% CAS No.: 67-64-1 Molecular Formula: C3H6O Formula Weight: 58.08 Flash Point: -17°(1°F) Refractive Index: 1.3590 Odor: Characteristic Melting Point: -94° Boiling Point: 56° Beilstein Reference: 635680 Merck Reference: 14,66 UN No.: 1090 MDL No.: MFCD00008765

Synonyms:  2-Propanone, DMK, Propanone, Dimethyl ketone
  • CAS Number:67-64-1
  • Formula:CH₃COCH₃
  • MDL Number:MFCD00008765
  • Molecular Weight:58.08 g/mol
  • Merck Index:14,66
  • Boiling Pt:56 °C
  • Melting Pt:–94 °C
  • Density:0.791
  • Flash Pt:–17 °C (1 °F)
  • Storage Temperature:Ambient
  • EINECS:200-662-2
  • UN:1090
  • ADR:3,II
  • REACH:01-2119471330-49
Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • CAAAAL10407-AU
  • CAAAAL10407-0F
  • Environmentally Preferable:
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Environmental certification:
  • Chem21 Solvent selection guide recommended after discussion
  • Chem21 Solvent selection guide recommended after discussion

Specifications

Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Bottle
    1 L
    Log in for availability
    Log in for availability
    Each (1l)
    Each (1l) - $56.42
    $56.42
    Each (1l)
       $56.42

    Supplier Number: L10407-AU
    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    CAAAAL10407-AU
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent selection guide recommended after discussion
      Safer Human and Environmental Health
  • Bottle
    2.5 L
    Log in for availability
    Log in for availability
    Each (3l)
    Each (3l) - $107.93
    $107.93
    Each (3l)
       $107.93

    Supplier Number: L10407-0F
    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    CAAAAL10407-0F
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent selection guide recommended after discussion
      Safer Human and Environmental Health

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.