Nunc® CryoPens, Thermo Scientific
Catalog # CA24270-010
Supplier: Thermo Fisher Scientific
Specifications
- Color:Black
- Description:Cryo pens
- Environmentally Preferable:
- Cat. no.:CA24270-010
- Supplier No.:343850
Specifications
About this item
Nunc® CryoPens come with indelible, quick-drying ink and extra fine tips
They are ideal for use with Nunc® Cryo Color Coders as part of a versatile and comprehensive filing system.